Learn More
Tocris Bioscience™ A 412997 dihydrochloride
Descripción
A 412997 dihydrochloride is a selective agonist for the dopamine D4 receptor (Ki values are 7.9 and 12.1 nM for human D4 and rat D4 receptors). Displays no affinity (<1000 nM) for other dopamine receptors. Shows efficacy in rat models of ADHD and short-term memory.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide |
| CAS | 1347744-96-0 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 5 mg |
| Fórmula molecular | C19H23N3O.2HCl |
| Formula Weight (peso de la fórmula) | Observed MW: 382.33 |