Learn More
Tocris Bioscience™ A 410099.1 amide-PEG2-amine
Descripción
A 410099.1 amide-PEG2-amine is a functionalized IAP ligand for PROTAC® research and development; incorporates an IAP ligand plus a short amide-PEG linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | tert-Butyl [-1-[[-2-[(2 S,4 S)-4-[2-[2-(2-aminoethoxy)ethoxy]acetamido]-2-[[(R)-1,2,3,4-tetrahydronaphthalen-1-yl]carbamoyl]pyrrolidin-1-yl]-1-cyclohexyl-2-oxoethyl]amino]-1-oxopropan-2-yl](methyl)carbamate |
| CAS | 2415256-16-3 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C38H60N6O8 |
| Pureza | 95% |