Learn More
A-196, MedChemExpress
Código de producto. 16469496
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16469496 50 mg 50 miligramos
16445587 5 mg 5 miligramos
16414256 100 mg 100 miligramos
16402848 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16469496 Proveedor Medchem Express N.º de proveedor HY100201
MedChemExpress A-196 is a potent and selective inhibitor of SUV420H1 and SUV420H2 with IC50 values of 25 nM and 144 nM, respectively. A-196 inhibits SUV4-20 biochemically in a substrate-competitive manner. A-196 represents a first-in-class chemical probe of SUV4-20 to investigate the role of histone methyltransferases in genomic integrity.
A-196 is a potent and selective inhibitor of SUV420H1 and SUV420H2 with IC50 values of 25 nM and 144 nM, respectively. A-196 inhibits SUV4-20 biochemically in a substrate-competitive manner. A-196 represents a first-in-class chemical probe of SUV4-20 to investigate the role of histone methyltransferases in genomic integrity.
Especificaciones
Nombre del producto químico o material A-196
Forma física Solid
CAS 1982372-88-2
Color Off-White
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C18H16Cl2N4
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 31 mg/mL (86.29 mM)
SMILES ClC1=C(Cl)C=C(C(C2=CC=NC=C2)=NN=C3NC4CCCC4)C3=C1
Peso molecular (g/mol) 359.25
Formula Weight (peso de la fórmula) 359.25
Porcentaje de pureza 99.61%