Learn More
Capreomycin sulfate, 90%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar J66684.03
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Thought to inhibit protein synthesis by inhibiting normal ribosomal functions
Especificaciones
| Capreomycin sulfate | |
| White | |
| >110°C (230°F) | |
| MFCD00079032 | |
| Freely soluble in water | |
| OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1 | |
| 766.79 | |
| 90% |
| 1405-37-4 | |
| 1g | |
| C25H46N14O12S | |
| 1377°C | |
| AJQVUIHGEOLMDY-SOCRLDLMNA-N | |
| 3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid | |
| 766.78 |