Learn More
Clindamycin phosphate, Thermo Scientific™
Antibiotic U-28508E, effective against Gram-positive bacteria
Marca: Thermo Scientific Alfa Aesar J63387.MC
| Cantidad | 100mg |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
| Clindamycin phosphate | |
| White | |
| C18H34ClN2O8PS | |
| 14,2356 | |
| UFUVLHLTWXBHGZ-AVENPWRCSA-N | |
| [(2R,3R,4S,5R,6R)-6-[(1S,2R)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate | |
| 71314904 |
| 24729-96-2 | |
| 100mg | |
| MFCD07793328 | |
| Freely soluble in water | |
| CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)OP(=O)(O)O)O)O)C(C)Cl | |
| 504.96 | |
| 504.96 |