Learn More
8-(dietilamino)octilo 3,4,5-trimetoxibenzoato hidrocloruro, 97 %, Thermo Scientific Chemicals
Código de producto. 15440247
Cambiar vista
Click to view available options
Cantidad:
50 mg
250 mg
Tamaño de la unidad:
250 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15440247 50 mg 50 miligramos
15430247 250 mg 250 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15440247 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J62528.MF
Solicitar formato a granel o personalizado
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Inhibe la movilización intracelular de calcio
Inhibits intracellular calcium mobilization
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 53464-72-5
Fórmula molecular C22H38ClNO5
Peso molecular (g/mol) 432
Número MDL MFCD00012502
Clave InChI KFJZVXKPPQIYCG-UHFFFAOYSA-N
Sinónimo 8-diethylamino octyl 3,4,5-trimethoxybenzoate hydrochloride, tmb-8 hydrochloride, tmb-8, hydrochloride, 8-diethylamino octyl-3,4,5-trimethoxybenzoate, 8-n,n-diethylamino-octyl-3,4,5-trimethoxybenzoate, hcl, 8-diethylamino octyl-3,4,5-trimethoxybenzoate hydrochloride, 3,4,5-trimethoxybenzoic acid 8-diethylamino octyl ester, hydrochloride, tmb 8 hydrochloride, acmc-1b069, regid_for_cid_6917843
PubChem CID 6917843
Nombre IUPAC 8-(dietilamino)octil 3,4,5-trimetoxibenzoato;clorhidrato
SMILES [H+].[Cl-].CCN(CC)CCCCCCCCOC(=O)C1=CC(OC)=C(OC)C(OC)=C1
Especificaciones
Nombre del producto químico o material 8-(Diethylamino)octyl 3,4,5-trimethoxybenzoate hydrochloride
Punto de fusión 92°C to 97°C
Color Blanco
Cantidad 50 mg
Formula Weight (peso de la fórmula) 432
Porcentaje de pureza 0.97
Forma física Polvo