Learn More
6bK, Tocris Bioscience™
Insulin degrading enzyme (IDE) inhibitor
Marca: Tocris Bioscience 5402/500U
Descripción
6bK is an insulin degrading enzyme (IDE) inhibitor (IC50 = 50 nM); regulates insulin, glucagon and amylin levels, and improves glucose tolerance in a diabetic mouse model.This compound is supplied in net weight as the TFA salt.
Especificaciones
| (3R,6S,9S,12E,16S)-9-(4-Aminobutyl)-3-[(4-benzoylphenyl)methyl]-6-(cyclohexylmethyl)-2,5,8,11,14-pentaoxo-1,4,7,10,15-pentaazacycloeicos-12-ene-16-carboxamide | |
| 1417537-93-9 | |
| >97% | |
| 500 μg |