Learn More
MedChemExpress 5-TAMRA
Código de producto. 16466918
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16466918 10 mg 10 miligramos
16452977 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16466918 Proveedor MedChemExpress N.º de proveedor HY15942
MedChemExpress 5-TAMRA (5-Carboxytetramethylrhodamine), SE and its conjugates yield bright, pH-insensitive orange-red fluorescence (approximate excitation/emission maxima ~546/579) with good photostability.
Especificaciones
Cantidad 10 mg
Peso molecular 430.45 g/mol
Solubilidad DMSO : ≥ 63 mg/mL (146.36 mM)
Contenido y almacenamiento 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Condiciones de envío Room temperature
CAS 91809-66-4