Learn More
5-Fluoro-5'-desoxiuridina, Thermo Scientific Chemicals
Código de producto. 16594354 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16594354 1 g 1 g
16504364 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16594354 Proveedor Thermo Scientific Acros N.º de proveedor 462790010
Solicitar formato a granel o personalizado
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 9-5-3094
Fórmula molecular C9H11FN2O5
Peso molecular (g/mol) 246.19
Número MDL MFCD00866530 MFCD00866530
Clave InChI ZWAOHEXOSAUJHY-ZIYNGMLESA-N
Nombre IUPAC 1-[(2R,3R,4S,5R)-3,4-dihidroxi-5-metiloxolan-2-il]-5-fluoro-1,2,3,4-tetrahidropirimidin-2,4-diona
SMILES C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(=O)NC1=O
Especificaciones
Nombre del producto químico o material 5-Fluoro-5 '-deoxyuridine
Punto de fusión 188.0°C to 192.0°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 246.19