Learn More
Tocris Bioscience™ 5-Amino-Thalidomide
Descripción
5-Amino-Thalidomide is a functionalized cereblon (CRBN) ligand for PROTAC® research and development; incorporates an E3 ligase ligand with an amine on the C5 position of the phthalimide ring for onward chemistry to generate Protein Degraders with rigid linkers. Degraders with modifications on the C5 position reduce off-target zinc-finger proteins degradation.
Part of a range of functionalized tool molecules for PROTAC® Degrader R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 5-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 191732-76-0 |
| Cantidad | 100 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C13H11N3O4 |
| Pureza | 0.95 |