Learn More
SHOWA™ 406 Double Latex-Coated Insulated Cold-Weather Gloves
Código de producto. 17747607
Change view
Click to view available options
Tamaño:
M
L
XL
2XL
Tamaño de la unidad:
5 pares
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño unitSize
17747607 XL 5 pares
17737607 M 5 pares
17727607 L 5 pares
17757607 2XL 5 pares
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 17747607 Proveedor SHOWA™ N.º de proveedor 40609XL
Solo quedan null
Añadir a la cesta
Edge
Añadir a la cesta
Water-repellant gloves are designed to remain dry when used with liquids, while intelligent acrylic insulation protects hands from freezing temperatures down to -30°C.
  • Double-dipped latex coating with a rough texture for enhanced grip capabilities
  • Intelligent acrylic insulation protects hands from temperatures down to -30°C
  • Insulation helps to reduce perspiration inside the glove to preserve grip and tactility
  • High visibility orange exterior increases user safety in low-light applications
  • Seamless knit construction for a snug fit to increase warmth and dexterity
Especificaciones
Grado de abrasión ANSI ANSI 2
Área revestida Palm
Estructura Seamless Knit
Coating Material Látex
Color Hi-vis Orange, Negro
Cut Resistance Rating ANSI A1
Acabado exterior Parcialmente texturizado
Hand Específico
Latex Free No
Material Nylon, Polyester
Tamaño XL
Para utilizar con (aplicación) Brick Laying, Outdoor Maintenance, Refrigeration, and Shipping/Receiving
Guantes con puño de color Yellow
Estilo de puño de los guantes Elastizado
Material de revestimiento Acrílico
Índice de pinchazos ANSI/ISEA 105-16 Level 1
Serie 406
Estilo de pulgar Straight
Longitud (imperial) 11 in