Learn More
Melford 4-Methylumbelliferyl-N-acetyl-B-D-glucosaminide
Código de producto. 30018498
Change view
Click to view available options
Cantidad:
100 mg
1 g
5 g
Tamaño de la unidad:
1 g
100 miligramos
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30018498 100 mg 100 miligramos
30018499 1 g 1 g
30018500 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30018498 Proveedor Melford N.º de proveedor M641000.1
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase.
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase. Ultrasensitive. Checked by actual enzymatic assay.