Learn More
4-Yodo-1H-pirazol, 99 %, Thermo Scientific Chemicals
Catalog Number 15412838
Click to view available options
Cantidad:
1 g
5 g
25 g
3469-69-0
C3H3IN2
193.975
MFCD00005244
LLNQWPTUJJYTTE-UHFFFAOYSA-N
4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295
77022
CHEBI:45260
4-yodo-1H-pirazol
C1=C(C=NN1)I
3469-69-0
C3H3IN2
193.975
MFCD00005244
LLNQWPTUJJYTTE-UHFFFAOYSA-N
4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295
77022
CHEBI:45260
4-yodo-1H-pirazol
C1=C(C=NN1)I
4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Se utilizó 4-yodopirazol en una síntesis de heterobiales mediada por indio. Se somete a yodación en presencia de yodo e hidróxido amónico para producir 3,4-di-yodo- y 3,4,5-tri-yiodo-pirazol.
Solubilidad
Soluble en agua.
Notas
Sensible a la luz. Incompatible con los agentes oxidantes. Almacenar a temperatura ambiente.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Iodo-1H-pyrazole
Punto de fusión 108°C to 110°C
Cantidad 1 g
Sensibilidad Light sensitive
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 193.97
Porcentaje de pureza 99%