missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCRL3 Polyclonal Antibody

Descripción
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
Especificaciones
Especificaciones
| Antígeno | FCRL3 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | FCRL3 |
| N.º de referencia del gen | Q96P31 |
| Alias de gen | BXMAS1-like protein 2; CD307c; CD307e; Fc receptor homolog 3; Fc receptor like 3; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRL3; Fcrl5; fcR-like protein 3; FcR-like protein 5; hIFGP3; IFGP family protein 3; IFGP3; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; mBXMH2; moFcRH3; rCG62831-like; SH2 domain-containing phosphatase anchor protein 2; SPAP2 |
| Símbolos de los genes | FCRL3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?