Learn More
4-Cloro-7-fluoro-2-metilquinolina, 95 %, Thermo Scientific Chemicals
Descripción
4-Chloro-7-fluoro-2-methylquinoline is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
La 4-cloro-7-fluoro-2-metilquinolina se utiliza como intermedio farmacéuticos.
Notas
Incompatible con los agentes oxidantes. Almacenar en un área fresca, seca y bien ventilada.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Chloro-7-fluoro-2-methylquinoline |
| Cantidad | 1 g |
| Formula Weight (peso de la fórmula) | 195.63 |
| Porcentaje de pureza | 95% |