Learn More
4'-Cloro-3'-fluoroacetofenona, 97 %, Thermo Scientific Chemicals
Descripción
4'-Chloro-3'-fluoroacetophenone is a halogenated benzene derivative used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
La 4′-cloro-3′-fluoroacetofenona es un derivado del benceno halogenado que se utiliza en síntesis orgánica.
Solubilidad
Ligeramente soluble en agua.
Notas
Incompatible con agentes oxidantes fuertes.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4'-Chloro-3'-fluoroacetophenone |
| Cantidad | 1 g |
| Información de solubilidad | Slightly soluble in water. |
| Formula Weight (peso de la fórmula) | 172.59 |
| Porcentaje de pureza | 97% |