Learn More
4-aminoazobenceno, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Aminoazobenzene |
| CAS | 60-09-3 |
| Fórmula molecular | C12 H11 N3 |
| Cantidad | 5 g |
| Sinónimo | Benzenamine, 4-(2-phenyldiazenyl)-, Benzenamine, 4-(phenylazo)- (9CI), C.I. Solvent Yellow 1 (7CI,8CI), 4-(2-Phenyldiazenyl)benzenamine, 4-(Phenylazo)aniline, 4-(Phenylazo)benzenamine, 4-(Phenyldiazenyl)aniline, 4-Aminoazobenzene, 4-Aminoazobenzol, Aniline yellow, Brasilazina Oil Yellow G, C.I. 11000, Cellitazol R, Ceres Yellow R, Fast Spirit Yellow, Fat Yellow AAB, NSC 2032, Oil Soluble Aniline Yellow, Oil Yellow AAB, Oil Yellow AB, Oil Yellow AN, Oil Yellow B, Organol Yellow 2A, Solvent Yellow 1, Solvent yellow, Somalia Yellow 2G, Sudan Yellow R, p-(Phenylazo)aniline, p-Aminoazobenzene, p-Aminoazobenzol, p-Aminodiphenylimide |
| Fórmula InChI | InChI=1S/C12H11N3/c13-10-6-8-12(9-7-10)15-14-11-4-2-1-3-5-11/h1-9H,13H2 |
| SMILES | Nc1ccc(cc1)N=Nc2ccccc2 |
| Nombre IUPAC | 4-[(E)-fenildiazenil]anilina |
| Peso molecular (g/mol) | 197.24 |
| Formula Weight (peso de la fórmula) | 197.0953 |