Learn More
3M™ 4510 Series Disposable Protective Coveralls
The 3M™ Protective Coverall 4510 is disposable safety work wear made of a high quality microporous laminated material. It helps provide basic barrier protection against light liquid splashes and hazardous dusts.
Marca:  3M™ 7000089662
Código de producto. 12952315
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
The key features make this protective coverall a great disposable PPE option. The 3M protective coverall 4510 helps protect against light liquid splashes and hazardous dusts. Typical applications for this safety work wear may include: Paint Spraying, Metal Polishing, Machine or Vehicle Maintenance (Oils and Greases), and General Industrial Clean-up and Processing.
Product recommended for the following industries:  Abatement/Remediation, Agriculture, Cleaning and Maintenance, General Industry/Manufacturing, Non-Hazardous Operations, Painting, Pharmeceutical Cleanroom, Welding/Metalwork Product recommended for the following industries: Abatement/Remediation, Agriculture, Cleaning and Maintenance, General Industry/Manufacturing, Non-Hazardous Operations, Painting, Pharmeceutical Cleanroom, Welding/Metalwork.
  • Protective coverall helps protect against certain light liquid splashes and hazardous dusts
  • Microporous PE laminate offers a barrier against hazardous particles and liquid splashes
  • Breathable, SMMMS fabric back panel helps improve air ventilation and helps reduce heat build-up
  • Reinforcement gusset between the legs to help provide increased durability during bending and squatting
  • Meets government standards for Type 6 Splashes, Type 5 Dust, Anti-Static and Nuclear protection
  • Elastic waist and ankles for convenience and freedom of movement.
  • Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
  • Three panel hood design helps provide a more comfortable fit and a sleek, low-profile style.