Learn More
(3Beta,20S)-Pregn-5-ene-3,17,20-triol, TRC
Product Code. 30410123
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad
30410123 5 mg
30416080 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30410123 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP7123255MG
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 903-67-3
Fórmula molecular C21H34O3
Peso molecular (g/mol) 334.49
Sinónimo Pregn-5-ene-3beta,17,20alpha-triol, (3beta,17alpha,20S)-Pregn-5-ene-3,17,20-triol, Pregn-5-ene-3beta,17alpha,20alpha-triol, delta5-Pregnene-3beta,17alpha,20(S)-triol
Nombre IUPAC (3S,8R,9S,10R,13S,14S,17R)-17-[(1S)-1-hidroxietilo]-10,13-dimetil-1,2,3,4,7,8,9,11,12,14,15,16-dodecahidrociclopenta[a]fenantrino-3,17-diol
SMILES C[C@H](O)[C@@]1(O)CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Nombre del producto químico o material (3beta,20S)-Pregn-5-ene-3,17,20-triol
Cantidad 5 mg
Fórmula InChI InChI=1S/C21H34O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h4,13,15-18,22-24H,5-12H2,1-3H3/t13-,15-,16+,17-,18-,19-,20-,21-/m0/s1
Formula Weight (peso de la fórmula) 334.2508