missing translation for 'onlineSavingsMsg'
Learn More

Labconco 3 ft. Nexus Horizontal Clean Bench with 21 in. Depth, Standard Interior and Base Stand, 230 V SC

Código de producto. 18897075 Tienda Labconco Corporation Productos
Cambiar vista
Click to view available options
Tensión:
230 V
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tensión Tamaño de la unidad
18897075 230 V cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18897075 Proveedor Labconco N.º de proveedor 5330530
 Artículo restringido
Fecha estimada de envoi: 15-02-2027
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment, large, 5 in. touchscreen interface features Nexus OS™ software simplifies operation, maintains records and provides user customizaton of system

The Nexus provides an ISO Class 4 clean airflow environment for protection of critical, non-hazardous samples.

Laminar airflow is driven by an ECM blower powered with Constant Airflow Profile (CAP™) technology for continuous, precise airflow that is energy efficient with long-lasting HEPA filters.

The sleek Nexus Horizontal Clean Bench design includes tempered safety glass and a stainless-steel interior option for effortless cleaning and durability in critical environments.

The touchscreen interface features Nexus OS™ software that simplifies operation, provides access to user settings and displays detailed alert messages when needed. Operational logs and maintenance records are stored onboard and can be accessed remotely for worry-free compliance management.

  • Fully scannable 99.99+% supply HEPA filter, 99.999% efficient ULPA filter available
  • Electronically Commutated Motor (ECM)
  • Unidirectional, laminar airflow
  • Constant Airflow Profile™ (CAP) airflow monitoring system
  • Nexus Operating System and full color, 5 in. touchscreen featuring:
    • Simplified home screen for easy start up and shut down operations
    • Night-Smart™ for idling the blower when the system isn't in use
    • Full operational, maintenance and certification logs for detailed record keeping
    • Numeric and visual HEPA Filter Life graphic for easy HEPA filter monitoring
    • Password security lock (optional activation) that requires user or admin password to operate the cabinet
    • Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 10% before alarming at 0%
    • Interval/elapsed timer for experiment monitoring
    • UV light timer (on models with UV)
    • 8 languages
  • Glass side panels
  • Spill-Stopper design protects HEPA filter from spills
  • USB port for data record export
  • Remote connectivity via Ethernet port
  • Prefilters to help extend HEPA filter life
  • Bright, glare-free LED lighting
  • Tempered safety glass and stainless steel side wall with one utility pass-through port and two service fixture punch-outs
  • Electrical Outlet(s)-Ceiling mount, Flat Stainless Steel Cover
    • R side on 3 and 4 ft. Models, R and L side on 5 to 8 ft. Models
    • 115 V GFCI, double receptacle, maximum amperage draw 5 A
    • 230 V, single receptacle, maximum amperage draw 3 A
  • 30.2 in. interior height
  • Optic White epoxy-coated steel
  • Nominal face velocity 85 fpm (0.43 m/s)
  • 10 ft. (3 m) power cord with plug
  • Five year warranty

Especificaciones

Tipo de producto Horizontal Clean Bench
Recubrimiento Epoxy
Tipo de filtro HEPA
Tipo de iluminación LED
Material Epoxy-coated Steel
Línea de productos Nexus
Fondo (imperial) 21 in.
Fondo (métrico) 533.4 mm
Altura (imperial) interior 30.2 in.
Altura (métrico) interior 767.08 mm
Clasificación IP None
Tensión 230 V
Anchura (imperial) 3 ft.
Anchura (métrico) 0.9144 m
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.