missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzoyl chloride, 96%, Thermo Scientific™

Product Code. 11426696
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11426696 1 g 1g
11436696 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11426696

Brand: Thermo Scientific Alfa Aesar B23849.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85345-76-2
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 193.00
MDL Number MFCD01631548
InChI Key VBYJOUAMJITBNL-UHFFFAOYSA-N
Synonym benzoyl chloride, 3-chloro-2-fluoro, buttpark 30\01-51, 3-chloro-2-fluoro benzoyl chloride, 3-chloro-2-fluoro-benzoyl chloride, benzoylchloride, 3-chloro-2-fluoro, 2-fluoro-3-chlorobenzoyl chloride, benzoyl chloride, 3-chloro-2-fluoro-9ci, 3-chloro-2-fluorobenzoylchloride, ksc451a3f, 2-fluoro-3-chlorobenzoylchloride
PubChem CID 2773598
IUPAC Name 3-chloro-2-fluorobenzoyl chloride
SMILES FC1=C(Cl)C=CC=C1C(Cl)=O

Specifications

Density 1.451
Boiling Point 202°C to 203°C
Flash Point 54°C (130°F)
Refractive Index 1.555
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.