Learn More
Alfa Aesar™ 3-Bromothiophene-2-carboxaldehyde, 94%
Código de producto. 11388666
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11388666 1 g 1 g
11398666 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11388666 Proveedor Alfa Aesar™ N.º de proveedor L19386.03
Solicitar formato a granel o personalizado
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Applications
3-Bromothiophene-2-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible in water.
Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
CAS 930-96-1
Fórmula molecular C5H3BrOS
Peso molecular (g/mol) 191.04
Número MDL MFCD00126680
Clave InChI BCZHCWCOQDRYGS-UHFFFAOYSA-N
Sinónimo 3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
PubChem CID 2797079
Nombre IUPAC 3-bromotiofeno-2-carbaldehído
SMILES BrC1=C(SC=C1)C=O
Especificaciones
Nombre del producto químico o material 3-Bromothiophene-2-carboxaldehyde
Punto de ebullición 110°C to 113°C (10 mmHg)
Cantidad 1 g
Beilstein 109757
Formula Weight (peso de la fórmula) 191.05
Porcentaje de pureza 94%