Learn More
3-[2-(etilhexil)oxilo]-1,2-propandiol, TRC
Product Code. 30133446
Change view
Click to view available options
Cantidad:
100 mg
1 g
10 g
Unit Size:
1 g
10 g
100 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30133446 10 g 10 g
30133442 1 g 1 g
30011515 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133446 Supplier Toronto Research Chemicals Supplier No. TRCE91877010G
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70445-33-9
Fórmula molecular C11 H24 O3
Peso molecular (g/mol) 204.31
Sinónimo 3-[(2-Ethylhexyl)oxy]-1,2-propanediol, 1-(2-Ethylhexyl) glycerol ether, Adekanol GE-RF, Ethylhexyl glycerin, Ethylhexylglycerin, Glycerol α-(2-ethylhexyl) ether, Glyceryl 1-mono(2-ethylhexyl) ether, Lexgard E, MinaCare Hexcine, Saskine 50, Sensiva AC 50JP, Sensiva SC 50, Sensiva SC 50JP, ethylhexylglycerine, 3-(2-Ethylhexyloxy)propane-1,2-diol
Nombre IUPAC 3-(2-etilhexoxi)propano-1,2-diol
SMILES CCCCC(CC)COCC(O)CO
Specifications
Nombre del producto químico o material 3-[2-(Ethylhexyl)oxyl]-1,2-propandiol
Notas de grado de pureza GC
Cantidad 10 g
Fórmula InChI InChI=1S/C11H24O3/c1-3-5-6-10(4-2)8-14-9-11(13)7-12/h10-13H,3-9H2,1-2H3
Formula Weight (peso de la fórmula) 204.1725
Porcentaje de pureza >95%