Learn More
Tocris Bioscience™ 27-Hydroxycholesterol
LXR agonist; also ER partial agonist
Marca: Tocris Bioscience™ 3907/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
27-Hydroxycholesterol is a liver X receptor (LXR) agonist. Also estrogen receptor partial agonist. Suppresses β-estradiol -mediated breast cancer cell proliferation in vitro. Also increases α-synuclein levels and induces apoptosis in human neuroblastoma cells in vitro. Endogenous cholesterol metabolite.Especificaciones
| (3β,25R)-Cholest-5-ene-3,26-diol | |
| 20380-11-4 | |
| 27 OHC, 27 HC | |
| 10 mg |