Learn More
Tocris Bioscience™ 25-Hydroxycholesterol
Antiviral; also α-crystallin (cryAB) chaperone
Marca: Tocris Bioscience™ 5741/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
25-Hydroxycholesterol is an antiviral. Stimulates expression of miR-185 microRNA which inhibits the hepatitis C virus life cycle via inhibition of cholesterol biosynthesis and virus cell membrane fusion. Also α-crystallin (cryAB) chaperone (Kd = 10.1 μM). Blocks α-crystallin aggregation and partially reverses cataract formation by binding and stabilizing soluble forms of cryAB in vitro and in vivo.Especificaciones
| Cholest-5-ene-3β,25-diol | |
| 2140-46-7 | |
| 402.65 | |
| Antiviral Compounds |
| Antiviral, Crystalline (cryAB) Chaperone | |
| C27H46O2 | |
| Store at -20°C | |
| 10 mg |
Título del producto
For Research Use Only