Learn More
Clorhidrato de 2-[(RS)-(4-metilfenil)fenilmetoxi]-N,N-dimetiletanamina, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride (4-Methyldiphenhydramine Hydrochloride) |
| CAS | 4024-34-4 |
| Fórmula molecular | C18 H23 N O . Cl H |
| Cantidad | 1 mg |
| Sinónimo | 4-Methyldiphenhydramine Hydrochloride, (RS)-N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamine Hydrochloride, N,N-Dimethyl-2-[(RS)-(4-methylphenyl)(phenyl)methoxy]ethanamine Hydrochloride, 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. F (EP) as Hydrochloride, para-Methylbenzyl Isomer Hydrochloride of Orphenadrine, Dimenhydrinate Imp. G (EP) as Hydrochloride, Diphenhydramine Hydrochloride Imp. B (EP) as Hydrochloride |
| Fórmula InChI | InChI=1S/C18H23NO.ClH/c1-15-9-11-17(12-10-15)18(20-14-13-19(2)3)16-7-5-4-6-8-16;/h4-12,18H,13-14H2,1-3H3;1H |
| SMILES | Cl.CN(C)CCOC(c1ccccc1)c2ccc(C)cc2 |
| Nombre IUPAC | N,N-dimetil-2-[(4-metilfenil)-fenilmetoxi]etanamina;clorhidrato |
| Peso molecular (g/mol) | 305.84 |
| Formula Weight (peso de la fórmula) | 305.1546 |