Learn More
2-Etoxipiridina-3-carboxaldehído, 97 %, Thermo Scientific Chemicals
Descripción
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Se utiliza como intermedio farmacéutico.
Notas
Sensible al aire. Mantener el recipiente herméticamente cerrado. Almacenar en un lugar fresco y seco y en recipientes bien sellados.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-Ethoxypyridine-3-carboxaldehyde |
| Cantidad | 1 g |
| Sensibilidad | Air Sensitive |
| Formula Weight (peso de la fórmula) | 151.17 |
| Porcentaje de pureza | 97% |