missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-di-tert-butylphenol, 97%

Product Code. 11450733
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11450733

Brand: Thermo Scientific Alfa Aesar A13218.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20834-61-1 | C14H21BrO | 285.225 g/mol

It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20834-61-1
Molecular Formula C14H21BrO
Molecular Weight (g/mol) 285.225
MDL Number MFCD00051597
InChI Key DIWZVAHZEOFSLS-UHFFFAOYSA-N
Synonym 2-bromo-4,6-di-tert-butylphenol, 2-bromo-4,6-di-t-butylphenol, 2,4-di-tert-butyl-6-bromophenol, phenol, 2-bromo-4,6-bis 1,1-dimethylethyl, phenol,2-bromo-4,6-bis 1,1-dimethylethyl, acmc-1crlp, 4,6-di-t-butyl-2-bromophenol, 6-bromo-2,4-di-t-butylphenol, 2-bromo4,6-di-tert-butylphenol, 2-bromo-4, 6-di-tert-butylphenol
PubChem CID 519822
IUPAC Name 2-bromo-4,6-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C(=C1)Br)O)C(C)(C)C

Specifications

Melting Point 55°C to 59°C
Quantity 100 g
Beilstein 2100150
Formula Weight 285.23
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-di-tert-butylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.