missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoic acid, 97%, Thermo Scientific™

Código de producto. 10525564
Cambiar vista
Click to view available options
Cantidad:
10 g
25 g
Tamaño de la unidad:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10525564 25 g 25 g
10152894 10 g 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10525564 Proveedor Thermo Scientific Maybridge N.º de proveedor SB01105EB
Solicitar formato a granel o personalizado

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Chemical Identifiers

CAS 385-00-2
Fórmula molecular C7H4F2O2
Peso molecular (g/mol) 158.104
Número MDL MFCD00002411
Clave InChI ONOTYLMNTZNAQZ-UHFFFAOYSA-N
Sinónimo benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
PubChem CID 9796
Nombre IUPAC ácido 2,6-difluorobenzoico
SMILES C1=CC(=C(C(=C1)F)C(=O)O)F

Specifications

Nombre del producto químico o material 2, 6-Difluorobenzoic acid
% mín. CAS 97.0
Intervalo de porcentaje de ensayo 97%
Fórmula lineal F2C6H3CO2H
Cantidad 25 g
Formula Weight (peso de la fórmula) 158.1
Porcentaje de pureza 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.