Learn More
2,4-dihidroxibenzofenona-d5, TRC
Product Code. 30375655
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30375655 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30375655 Supplier Toronto Research Chemicals Supplier No. TRCD45178310MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material 2,4-Dihydroxybenzophenone-d5
CAS 91586-06-0
Notas de grado de pureza HPLC
Fórmula molecular C13 D5 H5 O3
Cantidad 10 mg
Sinónimo 2,4-Dihydroxybenzophenone D5 (Phenyl D5)
Fórmula InChI InChI=1S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15H/i1D,2D,3D,4D,5D
SMILES [2H]c1c([2H])c([2H])c(C(=O)c2ccc(O)cc2O)c([2H])c1[2H]
Nombre IUPAC (2,4-dihidroxifenil)-(2,3,4,5,6-pentadeuteriofenil)metanona
Peso molecular (g/mol) 219.248
Formula Weight (peso de la fórmula) 219.094
Porcentaje de pureza >95