missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di-tert-butylphenol, 97%

Product Code. 11408427
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408427 250 g 250g
11498417 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408427 Supplier Thermo Scientific Alfa Aesar Supplier No. A18509.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-76-4 | C14H22O | 206.329 g/mol

2,4-Di-tert-butylphenol is used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts. It is a raw material for phosphite anti-oxidants. The antioxidants are primarily used in the manufacture of polyolefins. It is also used as an intermediate for the preparation of UV stabilizers and in manufacturing of pharmaceuticals and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Di-tert-butylphenol is used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts. It is a raw material for phosphite anti-oxidants. The antioxidants are primarily used in the manufacture of polyolefins. It is also used as an intermediate for the preparation of UV stabilizers and in manufacturing of pharmaceuticals and fragrances.

Solubility
Soluble in methanol, ether, acetone, chloroform. 2,4-Di-tert-butylphenol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-76-4
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.329
MDL Number MFCD00008828
InChI Key ICKWICRCANNIBI-UHFFFAOYSA-N
Synonym 2,4-di-tert-butylphenol, 2,4-di-t-butylphenol, phenol, 2,4-bis 1,1-dimethylethyl, antioxidant no. 33, 1-hydroxy-2,4-di-tert-butylbenzene, phenol, 2,4-di-tert-butyl, 2,4-bis tert-butyl phenol, prodox 146, prodox 146a-85x, unii-fob94g6hzt
PubChem CID 7311
IUPAC Name 2,4-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)C(C)(C)C

Specifications

Melting Point 53°C to 57°C
Density 0.887
Boiling Point 265°C
Flash Point 129°C (264°F)
Quantity 250 g
UN Number UN3077
Beilstein 1910383
Solubility Information Soluble in methanol,ether,acetone,chloroform. 2,4-Di-tert-butylphenol. Insoluble in water.
Formula Weight 206.33
Percent Purity 97%
Chemical Name or Material 2,4-Di-tert-butylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.