Learn More
Ácido 2-(2-yodofenil-d4)acético, TRC
Product Code. 30407679
Change view
Click to view available options
Cantidad:
100 mg
Unit Size:
100 miligramos
Product Code. Cantidad unitSize
30407679 100 mg 100 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30407679 Supplier Toronto Research Chemicals Supplier No. TRCI717532100MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material 2-(2-Iodophenyl-d4)acetic Acid
CAS 1217360-62-7
Fórmula molecular C8H3D4IO2
Cantidad 100 mg
Sinónimo (o-Iodophenyl-d4)acetic Acid, (2-Iodophenyl-d4)acetic Acid, 2-Iodobenzene-d4-acetic Acid
Fórmula InChI InChI=1S/C8H7IO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c(CC(=O)O)c(I)c1[2H]
Nombre IUPAC Ácido 2-(2,3,4,5-tetradeuterio-6-yodofenil)acético
Peso molecular (g/mol) 266.07
Formula Weight (peso de la fórmula) 265.9742