Learn More
17-DMAG hydrochloride, Tocris Bioscience™
Descripción
Water-soluble analog of 17-AAG (Cat. No. 1515) and geldanamycin (Cat. No. 1368). Binds the ATP binding site of Hsp90 and inhibits its chaperone activity. Displays more potent antitumor activity than 17-AAG (mean GI50 values are 53 and 123 nM for 17-DMAG and 17-AAG respectively).
Especificaciones
Especificaciones
| Nombre del producto químico o material | 17-DMAG hydrochloride |
| CAS | 467214-21-7 |
| Fórmula molecular | C32H49ClN4O8 |
| Cantidad | 1 mg |
| Sinónimo | 17-Demethoxy-17-[[2-(dimethylamino)ethyl]amino]geldanamycin hydrochloride |
| Clave InChI | DFSYBWLNYPEFJK-ZOKRJHFSSA-N |
| SMILES | CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)NCCN(C)C)C)OC)OC(=O)N)C)C)O)OC.Cl |
| Nombre IUPAC | [(3R,5S,6R,7S,8E,10S,11S)-21-[2-(dimethylamino)ethylamino]-6-hydroxy-5,11-dimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate;hydrochloride |
| Peso molecular (g/mol) | 653.214 |
| PubChem CID | 131954774 |
| Mostrar más |