missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiopronin, MedChemExpress
Descripción
Tiopronin is a diffusible antioxidant, an antidote to heavy metal poisoning and a radioprotective agent. Tiopronin can control the rate of cystine precipitation and excretion and has the potential for cystinuria, rheumatoid arthritis and hepatic disorders.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Tiopronin |
| Forma física | Solid |
| CAS | 2-2-1953 |
| Color | Blanco |
| Cantidad | 100 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C5H9NO3S |
| Para utilizar con (aplicación) | Metabolism-protein/nucleotide metabolism |
| Información de solubilidad | DMSO : ≥ 100 mg/mL (612.78 mM) ∣H2O : 100 mg/mL (612.78 mM; Need ultrasonic) |
| SMILES | O=C(O)CNC(C(S)C)=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?