Learn More
1-Methyladenosine, MedChemExpress
Código de producto. 16491027
Encompass_Preferred
Change view
Click to view available options
Cantidad:
50 mg
Tamaño de la unidad:
50 miligramos
16491027 50 mg 50 miligramos
1 options
Código de producto. 16491027
Marca: Medchem Express HY113081
MedChemExpress 1-Methyladenosine is an RNA modification originating essentially from two different reaction types, one catalyzed by enzymes and the other the result of the reaction of RNA with certain alkylating agents.
1-Methyladenosine is an RNA modification originating essentially from two different reaction types, one catalyzed by enzymes and the other the result of the reaction of RNA with certain alkylating agents.
Especificaciones
Nombre del producto químico o material 1-Methyladenosine
CAS 15763-06-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C11H15N5O4
Cantidad 50 mg
Información de solubilidad DMSO : 150 mg/mL (533.30 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN(C)C3=N)O1)O)O
Peso molecular (g/mol) 281.27
Formula Weight (peso de la fórmula) 281.27
Porcentaje de pureza 98.0%
Grado Research
Forma física Powder