Learn More
(1-etoxicarbonilciclopropil)trifenilfosfonio tetrafluoroborato, 98 %, Thermo Scientific Chemicals
Código de producto. 11463857
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
25 g
Tamaño de la unidad:
1 g
25 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
11463857 5 g 5 g
11453857 1 g 1 g
11473857 25 g 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 11463857 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A18305.06
Solicitar formato a granel o personalizado
CAS: 52186-89-7 | C24H24BF4O2P | 462.23 g/mol
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El (1-etoxicarbonilciclopropil)trifenilfosfonio tetrafluoroborato se utiliza como intermedio farmacéutico.
Solubilidad
Ligeramente soluble en agua.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
CAS 52186-89-7
Fórmula molecular C24H24BF4O2P
Peso molecular (g/mol) 462.23
Número MDL MFCD00051879
Clave InChI RGJYRMUGSAFITK-UHFFFAOYSA-N
Sinónimo 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborate
PubChem CID 11016017
SMILES F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Especificaciones
Nombre del producto químico o material (1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate
Punto de fusión 173°C to 179°C
Cantidad 5 g
Beilstein 4650673
Información de solubilidad Slightly soluble in water.
Nombre IUPAC [1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium; tetrafluoroboranuide
Formula Weight (peso de la fórmula) 462.23
Porcentaje de pureza 98%