Learn More
1,4-Diphenyl-1-butanone, 98%
Product Code. 11338346
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11338346 25 g 25g
11318346 1 g 1g
11328346 5 g 5g
3 options
This item is not returnable. View return policy
Product Code. 11338346
Brand: Thermo Scientific Alfa Aesar L15597.14
Request Bulk or Custom Format
This item is not returnable. View return policy
CAS: 5407-91-0 | C16H16O | 224.303 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5407-91-0
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
MDL Number MFCD00022005
InChI Key GBUMEGLMTNAXOM-UHFFFAOYSA-N
PubChem CID 79413
IUPAC Name 1,4-diphenylbutan-1-one
SMILES C1=CC=C(C=C1)CCCC(=O)C2=CC=CC=C2
Specifications
Melting Point 54°C to 56°C
Boiling Point 180°C (8 mmHg)
Quantity 25 g
Synonym 4-phenylbutyrophenone, 1,4-diphenyl-1-butanone, 1-butanone, 1,4-diphenyl, 4-phenylbutyrophenon, pubchem8921, 1-butanone,4-diphenyl, 1,4-diphenyl-butan-1-one, 1-butanone,1,4-diphenyl, 3-phenylpropyl phenyl ketone, acmc-1am65
Formula Weight 224.3
Percent Purity 98%
Chemical Name or Material 1,4-Diphenyl-1-butanone