Use el código promocional "25339" para aplicar la oferta.
Trietilaluminio, solución de 1,3 M en heptano, Thermo Scientific Chemicals
Use el código promocional "25339" para aplicar la oferta.
| Cantidad | 800 mL |
|---|
Identificadores de productos químicos
| CAS | 97-93-8 |
|---|---|
| Fórmula molecular | C6H15Al |
| Número MDL | MFCD00009015 |
| Clave InChI | VOITXYVAKOUIBA-UHFFFAOYSA-N |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| PubChem CID | 16682930 |
| Nombre IUPAC | trietilalumano |
| SMILES | CC[Al](CC)CC |
Specification
| Nombre del producto químico o material | Triethylaluminium |
|---|---|
| Color | Incolora |
| Nombre de nota | 1.3 M Solution in Heptane |
| CAS | 97-93-8 |
| CAS | 142-82-5 |
| Fórmula molecular | C6H15Al |
| Fórmula lineal | (C2H5)3Al |
| Número MDL | MFCD00009015 |
| Concentración | 19.5 to 20.5 wt% (on Al basis) |
| Densidad | 0.7300 g/mL |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Sinónimo | triethylalane,triethylaluminum solution, 25 wt. % in toluene,triethyl aluminum,triethyl-aluminum,aluminum, triethyl,triethylaluminium,triethylaluminum,triethylaluminum solution, 1.0 m in hexanes,triethyl-aluminum,unii-h426e9h3tt |
| Clave InChI | VOITXYVAKOUIBA-UHFFFAOYSA-N |
| SMILES | CC[Al](CC)CC |
| Nombre IUPAC | trietilalumano |
| Peso molecular (g/mol) | 114.17 |
| PubChem CID | 16682930 |
| Formula Weight (peso de la fórmula) | 114.17 |
| Envase | Frasco de vidrio |
| Cantidad | 800 mL |
| Forma física | Solución |
[Triethylaluminium, 1.M solution in heptane]
[Peligro]
[{code=AQUA CHRONIC 1, sequence=1, description=PELIGRO ACUÁTICO A LARGO PLAZO Categoría 1}, {code=AQUATIC ACUTE 1, sequence=2, description=PELIGRO ACUÁTICO AGUDO Categoría 1}, {code=ASP. TOX. 1, sequence=3, description=Toxicidad por aspiración Categoría 1}, {code=EYE DAM. 1, sequence=4, description=Lesión ocular grave/irritación ocular Categoría 1}, {code=FLAM. LIQ. 2, sequence=5, description=Líquido inflamable Categoría 2}, {code=PYR. LIQ. 1, sequence=6, description=Líquidos pirofóricos Categoría 1}, {code=SKIN CORR. 1B, sequence=7, description=Corrosión o irritación cutáneas Categoría 1B}, {code=STOT SE 3, sequence=8, description=Toxicidad específica para determinados órganos Categoría 3}, {code=WATER-REACT. 1, sequence=9, description=Sustancia que\, en contacto con el agua\, desprende gases inflamables Categoría 1}]
[{code=H304, sequence=1, description=Puede ser mortal en caso de ingestión y penetración en las vías respiratorias.}, {code=H318, sequence=2, description=Provoca lesiones oculares graves.}, {code=H400, sequence=3, description=Muy tóxico para los organismos acuáticos.}, {code=H410, sequence=4, description=Muy tóxico para los organismos acuáticos, con efectos nocivos duraderos.}, {code=H314, sequence=5, description=Provoca quemaduras graves en la piel y lesiones oculares graves.}, {code=H336, sequence=6, description=Puede provocar somnolencia o vértigo.}, {code=H250, sequence=7, description=Se inflama espontáneamente en contacto con el aire.}, {code=H260, sequence=8, description=En contacto con el agua desprende gases inflamables que pueden inflamarse espontáneamente.}, {code=H225, sequence=9, description=Líquido y vapores muy inflamables.}, {code=EUH014, sequence=91, description=Reacciona violentamente con el agua.}]
[{code=P210, sequence=50, description=Mantener alejado del calor, de superficies calientes, de chispas, de llamas abiertas y de cualquier otra fuente de ignición. No fumar.}, {code=P231+P232, sequence=51, description=Manipular en gas inerte. Proteger de la humedad.}, {code=P280, sequence=52, description=Llevar guantes/prendas/gafas/máscara de protección.}, {code=P301+P330+P331, sequence=53, description=EN CASO DE INGESTIÓN: Enjuagarse la boca. NO provocar el vómito.}, {code=P303+P361+P353, sequence=54, description=EN CASO DE CONTACTO CON LA PIEL (o el pelo): Quitar inmediatamente todas las prendas contaminadas. Aclararse la piel con agua/ducharse.}, {code=P305+P351+P338, sequence=55, description=EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.}, {code=P310, sequence=56, description=Llamar inmediatamente a un CENTRO DE TOXICOLOG A/médico/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS07, sequence=3, description=EXCLAMATION MARK}, {code=GHS08, sequence=4, description=HEALTH HAZARD}, {code=GHS09, sequence=5, description=ENVIRONMENT}]
Identificador del producto
- Triethylaluminium, 1.M solution in heptane
Indicación de advertencia
- Peligro
Categoría de peligro
- PELIGRO ACUÁTICO A LARGO PLAZO Categoría 1
- PELIGRO ACUÁTICO AGUDO Categoría 1
- Toxicidad por aspiración Categoría 1
- Lesión ocular grave/irritación ocular Categoría 1
- Líquido inflamable Categoría 2
- Líquidos pirofóricos Categoría 1
- Corrosión o irritación cutáneas Categoría 1B
- Toxicidad específica para determinados órganos Categoría 3
- Sustancia que\, en contacto con el agua\, desprende gases inflamables Categoría 1
Indicación de peligro
- H304- Puede ser mortal en caso de ingestión y penetración en las vías respiratorias.
- H318- Provoca lesiones oculares graves.
- H400- Muy tóxico para los organismos acuáticos.
- H410- Muy tóxico para los organismos acuáticos, con efectos nocivos duraderos.
- H314- Provoca quemaduras graves en la piel y lesiones oculares graves.
- H336- Puede provocar somnolencia o vértigo.
- H250- Se inflama espontáneamente en contacto con el aire.
- H260- En contacto con el agua desprende gases inflamables que pueden inflamarse espontáneamente.
- H225- Líquido y vapores muy inflamables.
- EUH014- Reacciona violentamente con el agua.
Consejos de prudencia
- P210- Mantener alejado del calor, de superficies calientes, de chispas, de llamas abiertas y de cualquier otra fuente de ignición. No fumar.
- P231+P232- Manipular en gas inerte. Proteger de la humedad.
- P280- Llevar guantes/prendas/gafas/máscara de protección.
- P301+P330+P331- EN CASO DE INGESTIÓN: Enjuagarse la boca. NO provocar el vómito.
- P303+P361+P353- EN CASO DE CONTACTO CON LA PIEL (o el pelo): Quitar inmediatamente todas las prendas contaminadas. Aclararse la piel con agua/ducharse.
- P305+P351+P338- EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.
- P310- Llamar inmediatamente a un CENTRO DE TOXICOLOG A/médico/.