CAS RN 959860-85-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
cFMS Receptor Inhibitor II, MedChemExpress
MedChemExpress cFMS Receptor Inhibitor II is a CSF1R kinase inhibitor. CSF-1 is a cytokine.