CAS RN 957230-65-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Isosofagomina D-Tartrato, TRC
CAS: 957230-65-8 Fórmula molecular: C6 H13 N O3 . C4 H6 O6 Peso molecular (g/mol): 297.26 Nombre IUPAC: (2S,3S)-2,3-ácido dihidroxibutanedioico; (3R,4R,5R)-5-(hidroximetil)piperidina-3,4-diol SMILES: OC[C@H]1CNC[C@@H](O)[C@@H]1O.O[C@@H]([C@H](O)C(=O)O)C(=O)O
2
Afegostat D-Tartrate, MedChemExpress
MedChemExpress Afegostat D-Tartrate is a pharmacological chaperone, which specifically and reversibly binds acid-β-glucosidase (GCase) in the endoplasmic reticulum (ER) with high affinity.