CAS RN 957217-65-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BTZ043 Racemate, MedChemExpress
MedChemExpress BTZ043 Racemate (BTZ10526038) is the racemate of BTZ043. BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate.