CAS RN 956590-23-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
UNBS5162, MedChemExpress
MedChemExpress UNBS5162 is a pan-antagonist of CXCL chemokine expression, with anti-tumor activity.