CAS RN 944795-06-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Lck inhibitor 2, MedChemExpress
MedChemExpress Lck inhibitor 2 is a bis-anilinopyrimidine inhibitor of tyrosine kinases including LCK, BTK, LYN, SYK, and TXK. The IC50 values are 13nM, 9nM, 3nM, 26nM and 2nM for Lck, Btk, Lyn, Btk and Txk respectively