CAS RN 944261-79-4
IUPAC Nombre:
5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
Sinónimos:
5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
Peso molecular (g/mol):
357.79
Fórmula molecular:
C19H16ClNO4
InChi Key:
VHKBTPQDHDSBSP-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=CC(NC(=O)C2=CC=C(O2)C2=CC=C(Cl)C=C2)=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ A 803467
CAS: 944261-79-4 Fórmula molecular: C19H16ClNO4
2
A-803467, MedChemExpress
MedChemExpress A-803467 is a potent and selective tetrodotoxin-resistant Nav1.8 sodium channel blocker (IC50=8 nM). A-803467 has shown significant anti-nociception in neuropathic and inflammatory pain models. A-803467 enhances the chemosensitivity of conventional anticancer agents through interaction with the ATP-binding cassette subfamily G member 2 (ABCG2) transporter.