CAS RN 943298-08-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress CY7
MedChemExpress Cy7 (Sulfo-Cyanine7) is a fluorescence labeling agent (Ex=750 nm, Em=773 nm). Cyanine dyes are used to label proteins, antibodies, peptides, and oligonucleotides.