CAS RN 942999-61-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DGAT-1 inhibitor 2, MedChemExpress
MedChemExpress DGAT-1 inhibitor 2 is an effective inhibitor of DGAT-1;antiobesity agents.