CAS RN 934662-91-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Cobimetinib racemate, MedChemExpress
MedChemExpress Cobimetinib racemate (GDC-0973 racemate; XL518 racemate) is the racemate of Cobimetinib. Cobimetinib is a potent and selective MEK inhibitor.