CAS RN 928333-30-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IRAK inhibitor 2, MedChemExpress
MedChemExpress IRAK inhibitor 2 is interleukin-1 receptor associated kinase inhibitor .