CAS RN 927822-86-4
IUPAC Nombre:
2,5-dichloro-N-(2-{[2-(2,5-dichlorobenzenesulfonamido)ethyl]disulfanyl}ethyl)benzene-1-sulfonamide
Sinónimos:
2,5-dichloro-N-(2-{[2-(2,5-dichlorobenzenesulfonamido)ethyl]disulfanyl}ethyl)benzenesulfonamide
Peso molecular (g/mol):
570.35
Fórmula molecular:
C16H16Cl4N2O4S4
InChi Key:
REQLACDIZMLXIC-UHFFFAOYSA-N
SMILES:
ClC1=CC(=C(Cl)C=C1)S(=O)(=O)NCCSSCCNS(=O)(=O)C1=C(Cl)C=CC(Cl)=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ KC7F2
CAS: 927822-86-4 Fórmula molecular: C16H16Cl4N2O4S4
2
KC7F2, MedChemExpress
MedChemExpress KC7F2 is a potent hypoxia inducible factor-1 (HIF-1) pathway inhibitor with an IC50 of 20 μM in LN229-HRE-AP cells, and with potential as a cancer therapy agent.