CAS RN 925434-55-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ SRT 1720
Selective SIRT1 activator; induces autophagy and mitochondrial biogenesis
2
SRT 1720, MedChemExpress
MedChemExpress SRT 1720 is a selective activator of human SIRT1 with an EC1.5 of 0.16 μM, and shows less potent activities agaiinst SIRT2 and SIRT3 with EC1.5s of 37 μM and > 300 μM, respectively.